Clear polypropylene Bio-Tite 120ml 1 * 90 items
Catalog # SHAN591201
Supplier: SHANDON
New Product
Frequently Bought Together
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Phosphate Buffered Saline (PBS), pH 7,4
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals