Paraffin kit, Histoplast PE, 2 x 2.3kg W 1 * 2 items
Catalog # SHAN8330K
Supplier: SHANDON
New Product
Paraffin kit, Histoplast PE, 2 x 2.3kg W 1 * 2 items
Catalog # SHAN8330K
Supplier: SHANDON
New Product
Frequently Bought Together
Laboratory trays
Supplier: KARTELL
Acetone ≥99.9%, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
N,N-Dimethylacetamide 99.99-100.00%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
Measuring scoops, PP
Supplier: SEMADENI
KingFisher™ Deep Well Plates, Barcoded
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for HPLC, for UV spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cryoboxes, B41/B42/B61/B62/B72/B92-series
Supplier: GLW
Dimethyl sulphoxide 99.8+%, extra pure
Supplier: Thermo Fisher Scientific
Cryoboxes, coloured, Nalgene®
Supplier: Thermo Fisher Scientific
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific