Syntri Safeguard Pen-Fix,4x5L 1 * 4 items
Catalog # SHAN6115SS
Supplier: SHANDON
New Product
Frequently Bought Together
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Cryoboxes
Supplier: RATIOLAB
VWR® Electroporation Cuvettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Serological Pipettes, Open End
Supplier: VWR Collection