PARAFFIN TYPE 1 10X2LB 1 * 90 kg
Catalog # SHAN8334
Supplier: SHANDON
New Product
Frequently Bought Together
Dimethyl sulphoxide 99.9% for spectroscopy
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥85% (isomers basis, GC), Puriss.
Supplier: Honeywell Chemicals
2-Propanol ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
Water ACS
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.7%, pure
Supplier: Thermo Fisher Scientific
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals