0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
HiPur,Acetone,4x1GAL 1 * 4 items
SHAN9000-1
:
SHANDON
New Product
HiPur,Acetone,4x1GAL 1 * 4 items
SHAN9000-1
:
SHANDON
New Product
Frequently Bought Together
Reagents for Phosphate Determination using Lovibond® MD photometers
: Lovibond
Schiff reagent
: Thermo Fisher Scientific
Sodium hydroxide 10 N in aqueous solution, Biotech Reagent, J.T.Baker®
: Avantor
DPD NO.3 SOLUTION 1 * 100 mL
: Lovibond