ContactUsLinkComponent
Shipping case for calibration module CM96 consisting of: SP-Case 2 incl. ESD insert Internal dimensions: 195x140x40mm Foam: PE foam RG35 ESD black Foam insert made of PE foam RG35 ESD black Lid with PU foam ESD black, 5mm 1 * 1 items
Catalog # VWRI36912
Supplier:  VWR Collection
New Product
Shipping case for calibration module CM96 consisting of: SP-Case 2 incl. ESD insert Internal dimensions: 195x140x40mm Foam: PE foam RG35 ESD black Foam insert made of PE foam RG35 ESD black Lid with PU foam ESD black, 5mm 1 * 1 items
Catalog # VWRI36912
Supplier:  VWR Collection