ContactUsLinkComponent
,,14 components: 100ug/ml each of Al As Ca Cd Cr Cu Fe Hg K Mg Na Ni Pb Zn in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM110592L1
New Product
,,14 components: 100ug/ml each of Al As Ca Cd Cr Cu Fe Hg K Mg Na Ni Pb Zn in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM110592L1