Cy5 F-Luc mRNA (MRNA24C: is modified with N1-methylpseudouridine (N1-m) and labelled with Cy5 (Sulfo-Cyanine5)) 100 ug 1 * 100 G
Catalog # OZBIMRNA24C-100
Supplier: OZ BIOSCIENCES
New Product
Cy5 F-Luc mRNA (MRNA24C: is modified with N1-methylpseudouridine (N1-m) and labelled with Cy5 (Sulfo-Cyanine5)) 100 ug 1 * 100 G
Catalog # OZBIMRNA24C-100
Supplier: OZ BIOSCIENCES
New Product
Frequently Bought Together
Sample Spoons, Disposable, SteriWare®
Supplier: Sampling Systems
VWR®, Microcentrifuge Tubes with Flat Screw Caps
Supplier: VWR Collection
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Supplier: Corning
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Cryoboxes, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Test tubes
Supplier: KARTELL
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Stoppers for test tubes, Corning® Gosselin™
Supplier: Corning
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection