(S,S)-(4,5-DIHYDRO-4-ISOPROPYL-2-OXAZOLYL)-2-[DI(1-NAPHTHYL)PHOSPHINO]FERROCENE, 97% 1 * 500 mg
Catalog # ABCRAB426517-500MG
Supplier: abcr
(S,S)-(4,5-DIHYDRO-4-ISOPROPYL-2-OXAZOLYL)-2-[DI(1-NAPHTHYL)PHOSPHINO]FERROCENE, 97% 1 * 500 mg
Catalog # ABCRAB426517-500MG
Supplier: abcr
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
n-Pentane, EMSURE® for analysis, Supelco®
Supplier: Merck
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Tetrahydrofuran, anhydrous (max. 0.003% H₂O) ≥99.9% stabilised
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.8+%, extra pure
Supplier: Thermo Fisher Scientific