VIAL 343ML PP WHITE FLIP-TOP CDT-CLEAR 1 * 90 items
Catalog # 216-4906
Supplier: Capitol Europe
New Product
VIAL 343ML PP WHITE FLIP-TOP CDT-CLEAR 1 * 90 items
Catalog # 216-4906
Supplier: Capitol Europe
New Product
Frequently Bought Together
VWR® Facial Tissues
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
VWR® Disposable Spatulas, PP
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Dehydrated culture media
Supplier: Merck
ABSAUGSPITZE M6 ID/AD 2/2,9MM, L 151MM 1 * 1 ST
Supplier: METROHM
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Accessories for Cimarec+™ and SuperNuova+™ series
Supplier: Thermo Fisher Scientific