VIAL 50ML MLP W. FLIPTOP F. QUALITY DHIA 1 * 500 items
Catalog # 216-0876
Supplier: Capitol Europe
New Product
VIAL 50ML MLP W. FLIPTOP F. QUALITY DHIA 1 * 500 items
Catalog # 216-0876
Supplier: Capitol Europe
New Product
Frequently Bought Together
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Aspirator bottles with screw cap and spigot
Supplier: KARTELL
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
Hydrogen peroxide 6% (w/v) (20 volumes), GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Sterilin® Dippas® Milk Dippers
Supplier: Sterilin