ContactUsLinkComponent
This Uric Acid Assay Kit is a quantitative assay kit for measuring Uric Acid in serum, plasma, urine, and animal tissues. 1 * 48 Tests
Catalog # ANTIA319754-48
Supplier:  ANTIBODIES.COM
New Product
This Uric Acid Assay Kit is a quantitative assay kit for measuring Uric Acid in serum, plasma, urine, and animal tissues. 1 * 48 Tests
Catalog # ANTIA319754-48
Supplier:  ANTIBODIES.COM