ContactUsLinkComponent
This Urease Activity Assay Kit is a quantitative assay kit for measuring Urease activity in animal and plant tissue, cell, bacteria, serum, and plasma. 1 * 48 Tests
Catalog # ANTIA319753-48
Supplier:  ANTIBODIES.COM
New Product
This Urease Activity Assay Kit is a quantitative assay kit for measuring Urease activity in animal and plant tissue, cell, bacteria, serum, and plasma. 1 * 48 Tests
Catalog # ANTIA319753-48
Supplier:  ANTIBODIES.COM