ContactUsLinkComponent
This Triglyceride Assay Kit is a quantitative assay kit for measuring Triglyceride in serum, plasma, animal and plant tissues, cells , cell supernatant, and bacteria. 1 * 48 Tests
Catalog # ANTIA319752-48
Supplier:  ANTIBODIES.COM
New Product
This Triglyceride Assay Kit is a quantitative assay kit for measuring Triglyceride in serum, plasma, animal and plant tissues, cells , cell supernatant, and bacteria. 1 * 48 Tests
Catalog # ANTIA319752-48
Supplier:  ANTIBODIES.COM