ContactUsLinkComponent
This Total Glutathione Assay Kit is a quantitative assay kit for measuring Total Glutathione in animal and plant tissues, bacteria, cells, and serum. 1 * 96 Tests
Catalog # ANTIA319747-96
Supplier:  ANTIBODIES.COM
New Product
This Total Glutathione Assay Kit is a quantitative assay kit for measuring Total Glutathione in animal and plant tissues, bacteria, cells, and serum. 1 * 96 Tests
Catalog # ANTIA319747-96
Supplier:  ANTIBODIES.COM