ContactUsLinkComponent
This Reducing Sugar Assay Kit is a quantitative assay kit for measuring Reducing sugars in plant tissues, animal tissues, cells, bac teria, serum, and plasma. 1 * 48 Tests
Catalog # ANTIA319729-48
Supplier:  ANTIBODIES.COM
New Product
This Reducing Sugar Assay Kit is a quantitative assay kit for measuring Reducing sugars in plant tissues, animal tissues, cells, bac teria, serum, and plasma. 1 * 48 Tests
Catalog # ANTIA319729-48
Supplier:  ANTIBODIES.COM