ContactUsLinkComponent
This Alanine Aminotransferase Activity Assay Kit is a quantitative assay kit for measuring Alanine Aminotransferase activity in anim al tissues, plant tissues, cell culture (adherent or suspension cells), serum, and bacteria. 1 * 96 Tests
Catalog # ANTIA319658-96
Supplier:  ANTIBODIES.COM
New Product
This Alanine Aminotransferase Activity Assay Kit is a quantitative assay kit for measuring Alanine Aminotransferase activity in anim al tissues, plant tissues, cell culture (adherent or suspension cells), serum, and bacteria. 1 * 96 Tests
Catalog # ANTIA319658-96
Supplier:  ANTIBODIES.COM