This 6-Phosphofructokinase Activity Assay Kit is a quantitative assay kit for measuring 6-Phosphofructokinase activity in serum, pla sma, animal and plant tissues, cells, cell culture supernatant, and bacteria. 1 * 96 Tests
Catalog # ANTIA319652-96
Supplier: ANTIBODIES.COM