ContactUsLinkComponent
Mouse Apolipoprotein CIII ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Apolipoprotein CIII in serum, plasma, tissue homogenates, and other biological fluids. 1 * 96 Tests
Catalog # ANTIA319391-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Apolipoprotein CIII ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Apolipoprotein CIII in serum, plasma, tissue homogenates, and other biological fluids. 1 * 96 Tests
Catalog # ANTIA319391-96
Supplier:  ANTIBODIES.COM