p-Toluidine, 99% 1 * 250 g
Catalog # ABCRAB119205-250
Supplier: abcr
Frequently Bought Together
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Tetrachloroethylene ≥98%, TECHNICAL
Supplier: VWR Chemicals
Diethylene glycol for synthesis
Supplier: Merck
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8% for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Water, TraceSELECT™ for trace analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals