ContactUsLinkComponent
Lens Barrel Custom Probe. Custom Probe focusing lens barrel with the following specification: 316 SS, 200 mm length, Kalrez O-ring, Dia 1/4 inch 1 * 1 items
Catalog # ASYNASY-C-1105
Supplier:  ASYNT
New Product
Lens Barrel Custom Probe. Custom Probe focusing lens barrel with the following specification: 316 SS, 200 mm length, Kalrez O-ring, Dia 1/4 inch 1 * 1 items
Catalog # ASYNASY-C-1105
Supplier:  ASYNT