ContactUsLinkComponent
Novel Green Plus provides an easy 2-step method to stain DNAs band from DNA electrophoresis. This unique reagent ensures the DNA to be stained with a high sensitivity and good quality. Novel Green Plus is a next-generation DNA-binding dye ideal for u 1 * 500 µl
Catalog # ANTIA318984-500
Supplier:  ANTIBODIES.COM
New Product
Novel Green Plus provides an easy 2-step method to stain DNAs band from DNA electrophoresis. This unique reagent ensures the DNA to be stained with a high sensitivity and good quality. Novel Green Plus is a next-generation DNA-binding dye ideal for u 1 * 500 µl
Catalog # ANTIA318984-500
Supplier:  ANTIBODIES.COM