ContactUsLinkComponent
Novel Green provides an easy 2-step method to stain DNA bands from DNA electrophoresis. This unique reagent ensures the DNA to be st ained with a high sensitivity and good quality. Novel Green is a next-generation DNA-binding dye ideal for use in qua 1 * 500 µl
Catalog # ANTIA318983-500
Supplier:  ANTIBODIES.COM
New Product
Novel Green provides an easy 2-step method to stain DNA bands from DNA electrophoresis. This unique reagent ensures the DNA to be st ained with a high sensitivity and good quality. Novel Green is a next-generation DNA-binding dye ideal for use in qua 1 * 500 µl
Catalog # ANTIA318983-500
Supplier:  ANTIBODIES.COM