Pre-stained protein ladder. Covers the size range 10-180 kDa (Tris-glycine-SDS running buffer). Number of bands = 11. Single colour with enhanced intensity at approximately 25 kDa and approximately 72 kDa. Designed for monitoring SDS-PAGE separation 1 * 500 µl
Catalog # ANTIA318981-500
Supplier: Antibodies.com