ContactUsLinkComponent
Pre-stained protein ladder. Covers the size range 3.5-245 kDa (Tris-glycine-SDS running buffer). Number of bands = 13. Green band at approximately 25 kDa. Red band at approximately 75 kDa. Designed for monitoring SDS-PAGE separation and WB transfer w 1 * 500 µl
Catalog # ANTIA318980-500
Supplier:  ANTIBODIES.COM
New Product
Pre-stained protein ladder. Covers the size range 3.5-245 kDa (Tris-glycine-SDS running buffer). Number of bands = 13. Green band at approximately 25 kDa. Red band at approximately 75 kDa. Designed for monitoring SDS-PAGE separation and WB transfer w 1 * 500 µl
Catalog # ANTIA318980-500
Supplier:  ANTIBODIES.COM