ContactUsLinkComponent
Pre-stained protein ladder. Covers the size range 10-180 kDa (Tris-glycine-SDS running buffer). Number of bands = 10. Green band at approximately 25 kDa. Red band at approximately 75 kDa. Designed for monitoring SDS-PAGE separation and WB transfer ef 1 * 500 µl
Catalog # ANTIA318978-500
Supplier:  Antibodies.com
New Product
Pre-stained protein ladder. Covers the size range 10-180 kDa (Tris-glycine-SDS running buffer). Number of bands = 10. Green band at approximately 25 kDa. Red band at approximately 75 kDa. Designed for monitoring SDS-PAGE separation and WB transfer ef 1 * 500 µl
Catalog # ANTIA318978-500
Supplier:  Antibodies.com