ContactUsLinkComponent
pH-4 standard solution #100-4 material number: 3200043638 pH 4,01 buffer solution, 500 ml in dosing bottle NIST Traceable with certificate, accuracy +- 0,01 @ 25°C New 1 * 500 mL
Catalog # HORI90030016000
Supplier:  HORIBA
New Product
pH-4 standard solution #100-4 material number: 3200043638 pH 4,01 buffer solution, 500 ml in dosing bottle NIST Traceable with certificate, accuracy +- 0,01 @ 25°C New 1 * 500 mL
Catalog # HORI90030016000
Supplier:  HORIBA