ContactUsLinkComponent
,,20 components: p-Xylene [CAS:106-42-3] 2000mg/l Formic acid methyl ester [CAS:107-31-3] 3000mg/l Acetic acid-isopropyl ester [CAS:108-21-4] 3000mg/l m-Xylene [CAS:108-38-3] 2000mg/l Toluene [CAS:108-88-3] 4000mg/l Cyclohexane [CAS:110-82- 1 * 30 mL
Catalog # CMPLRD101282L03.6
New Product
,,20 components: p-Xylene [CAS:106-42-3] 2000mg/l Formic acid methyl ester [CAS:107-31-3] 3000mg/l Acetic acid-isopropyl ester [CAS:108-21-4] 3000mg/l m-Xylene [CAS:108-38-3] 2000mg/l Toluene [CAS:108-88-3] 4000mg/l Cyclohexane [CAS:110-82- 1 * 30 mL
Catalog # CMPLRD101282L03.6