ContactUsLinkComponent
,,9 components: Dimethoxymethane [CAS:109-87-5] 3700mg/l Propionaldehyde [CAS:123-38-6] 3000mg/l Butyraldehyde [CAS:123-72-8] 2600mg/l Mesityl oxide [CAS:141-79-7] 3000mg/l 2,3-Butanedione [CAS:431-03-8] 2000mg/l Acetaldehyde [CAS:75-07-0] 1 * 30 mL
Catalog # CMPLRD101281L03.6
New Product
,,9 components: Dimethoxymethane [CAS:109-87-5] 3700mg/l Propionaldehyde [CAS:123-38-6] 3000mg/l Butyraldehyde [CAS:123-72-8] 2600mg/l Mesityl oxide [CAS:141-79-7] 3000mg/l 2,3-Butanedione [CAS:431-03-8] 2000mg/l Acetaldehyde [CAS:75-07-0] 1 * 30 mL
Catalog # CMPLRD101281L03.6