Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Protein Concentration Service (tie-in addenda to custom conjugation service) 1 * 1 items
Catalog # AATBPC001
Supplier:
AAT Bioquest
New Product
Expand image
Protein Concentration Service (tie-in addenda to custom conjugation service) 1 * 1 items
Catalog # AATBPC001
Supplier:
AAT Bioquest
New Product
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Cell scrapers and cell lifters
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Cryoboxes with Dividers
Supplier: VWR Collection