CONDUCTIVITY STANDARD 100µS/CM 1 * 250 mL
Catalog # HAMI238934
Supplier: HAMILTON BONADUZ
New Product
CONDUCTIVITY STANDARD 100µS/CM 1 * 250 mL
Catalog # HAMI238934
Supplier: HAMILTON BONADUZ
New Product
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
CONDUCTIVITY STANDARD 1.3µS/CM 1 * 250 mL
Supplier: HAMILTON BONADUZ
Conductivity standards
Supplier: WTW
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Conductivity standards
Supplier: HAMILTON BONADUZ
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Cold Filter Plugging Point (CFPP) standards, Certified Reference Material
Supplier: VWR Chemicals
CONDUCTIVITY STANDARD 100µS/CM 1 * 250 mL
Supplier: HAMILTON BONADUZ
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals