ContactUsLinkComponent
High-grade precision thermometer, DIN 12778, solid stem, -10+480:2C, white backed, gallium filled, 450x6-7mm, total immersion, , durable pigment, with Works Certificate with 3 test points 1 * 1 items
Catalog # AMARL29006-GA/WC
Supplier:  Amarell
New Product
High-grade precision thermometer, DIN 12778, solid stem, -10+480:2C, white backed, gallium filled, 450x6-7mm, total immersion, , durable pigment, with Works Certificate with 3 test points 1 * 1 items
Catalog # AMARL29006-GA/WC
Supplier:  Amarell