Two interior electrical outlets UK, BS1363, rated 250V, 50/60Hz, 13A note: reduced temperature range of 15 C to 40 C with this op tion this option is factory installed, please order with Heratherm environmental chamber. 1 * 1 items
Catalog # 390-1719
Supplier: Thermo Fisher Scientific
Frequently Bought Together