Two interior electrical outlets EU, CEE7/4 (Schuko), rated 250V, 50/60Hz, 16A note: reduced temperature range of 15 C to 40 C wit h this option this option is factory installed, please order with Heratherm environmental chamber. 1 * 1 items
Catalog # 390-1718
Supplier: Thermo Fisher Scientific
Frequently Bought Together