HighPurS-CERT - Ammonium Chloride (NH4Cl) - min. 99.999%, ISO 17034 and ISO/IEC 17025 certified CRM, ISO 17034 and ISO 17025 Conservation 36 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 50 g
Catalog # CMPLPSAMCLC.50G
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together