ContactUsLinkComponent
,,ICP-MS Calibration Standard (XXI) - 29 components: 10mg/l each of Ag Al As Ba Be Bi Ca Cd Co Cr Cs Cu Fe Ga In K Li Mg Mn Na Ni Pb Rb Se Sr Tl U V Zn in HNO3 5% ,,CRM, ISO 17034 and ISO 17025, 1 * 500 mL
Catalog # CMPLRM001217L5
,,ICP-MS Calibration Standard (XXI) - 29 components: 10mg/l each of Ag Al As Ba Be Bi Ca Cd Co Cr Cs Cu Fe Ga In K Li Mg Mn Na Ni Pb Rb Se Sr Tl U V Zn in HNO3 5% ,,CRM, ISO 17034 and ISO 17025, 1 * 500 mL
Catalog # CMPLRM001217L5