Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Accessories for Corning CellSTACK culture chambers

Accessories for Corning CellSTACK culture chambers

Barbed cap with female MPC; Int. Ø: 9,525 mm; Tubing length: 152,4 mm
Katalogové číslo CORN11757
Dodavatel:   Corning
Accessories for Corning CellSTACK culture chambers
Katalogové číslo CORN11757
Dodavatel:  Corning
Accessories for Corning CellSTACK culture chambers
Accessories for Corning CellSTACK culture chambers

Barbed cap with female MPC; Int. Ø: 9,525 mm; Tubing length: 152,4 mm

Přejít na:
Price
13 220,00 Kč
Přihlásit se pro dostupnost
1 * 5 KS
Specifikace
  • Description:
    Barbed cap with female MPC; Int. Ø: 9,525 mm; Tubing length: 152,4 mm
  • Seq:
    2
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.