0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
COMPONENTS 8 MIX 1 * 2 mL
CMPLRD1087881.2
:
CUSTOM MADE CHEMICALS LAB
COMPONENTS 8 MIX 1 * 2 mL
CMPLRD1087881.2
:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Chinese ink, yellow, Q Path® stain for histology
: VWR Chemicals
Sulphide reagents 1 and 2
: Hach
Reagents for hydrazine determination using Lovibond® MD photometers
: Lovibond
Sodium hydroxide ≥98.0% (by acidimetry), pellets ACS, ISO for analysis
: PanReac AppliChem
Ferrozine mono-sodium salt hydrate
: Hach