HUMAN IGG4Κ ISOTYPE CONTROL, RECOMBINANT MONOCLONAL ANTIBODY (MOPC-21) 1 * 200 µG
Catalog # ENZOENZABS5850200
Supplier: Enzo Life Sciences
HUMAN IGG4Κ ISOTYPE CONTROL, RECOMBINANT MONOCLONAL ANTIBODY (MOPC-21) 1 * 200 µG
Catalog # ENZOENZABS5850200
Supplier: Enzo Life Sciences
Frequently Bought Together
Plugs for test tubes
Supplier: BOETTGER PAUL
Serological pipettes, Stripette™
Supplier: Corning
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes with Flat Screw Caps
Supplier: VWR Collection
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Microcentrifuge tubes, 0,5/1,5/2,0 ml
Supplier: Brand
Tricorn Empty Chromatography Columns
Supplier: Cytiva
Whatman™ Protran® BA85 & BA83 NC Western Blotting Membranes, Nitrocellulose
Supplier: Whatman products (Cytiva)
Spare Lid of Ice Pan 9L, colour option, RED, BLUE, GREEN, BLACK 1 * 1 items
Supplier: VWR Collection
VWR®, Cell Culture Insert Plates, PET
Supplier: VWR Collection
384 v well microplate, polypropylene, sterile 1 * 100 items
Supplier: Greiner Bio-One
VWR®, PCR/qPCR Plates, 96-Wells, Plastic
Supplier: VWR Collection
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Coolers for Reagent Reservoirs
Supplier: DIVERSIFIED BIOTECH
Electroporation cuvettes
Supplier: Thermo Fisher Scientific