CD3Ε RECOMBINANT MONOCLONAL ANTIBODY (500A2) (MOUSE IGG1Κ) 1 * 200 µG
Catalog # ENZOENZABS4280200
Supplier: ENZO LIFE SCIENCES
CD3Ε RECOMBINANT MONOCLONAL ANTIBODY (500A2) (MOUSE IGG1Κ) 1 * 200 µG
Catalog # ENZOENZABS4280200
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Transfer pipettes, Falcon®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
External thread cryogenic vials
Supplier: Corning
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
VWR®, Cryo Boxes
Supplier: VWR Collection
Storage tubes, MATRIX™
Supplier: THERMO MATRIX TECHNOLOGIES
Cell strainers, EASYstrainer™
Supplier: Greiner Bio-One
Blotting transfer papers
Supplier: Ahlstrom-Munksjö
VWR®, Cryoboxes, Half Size, 136×66,5 mm
Supplier: VWR Collection
Cryoboxes, 133x133 mm
Supplier: HEATHROW