AMPIPROBE BACTERIAL VAGINOSIS EXTRACTION CONTROLS 1 * 500 µl
Catalog # ENZOENZGEN2060500
Supplier: ENZO LIFE SCIENCES
AMPIPROBE BACTERIAL VAGINOSIS EXTRACTION CONTROLS 1 * 500 µl
Catalog # ENZOENZGEN2060500
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Potassium hydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydroxide 30% (w/w) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Test kits, cyanide, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide 97.0-100.5%, pellets NF, JP, Ph. Eur., BP
Supplier: VWR Chemicals
Test strips, tin, MQuant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, lead, Spectroquant®, Supelco®
Supplier: Merck
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
Test strips for glucose, MQuant®, Supelco®
Supplier: Merck
VWR® Cryolab Q Path, Freezing Aerosol
Supplier: VWR Chemicals
Ascorbic acid test strips, Dosatest
Supplier: VWR Chemicals
Ammonia solution ∼25% (NH₃ basis) in water, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond