0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
VISMODEGIB (FREE BASE) 1 * 100 mg
ENZOENZCHM2420100
:
Enzo Life Sciences
VISMODEGIB (FREE BASE) 1 * 100 mg
ENZOENZCHM2420100
:
Enzo Life Sciences
Frequently Bought Together
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Test strips for copper, MQuant®, Supelco®
: Merck
Test kits, nitrite, Spectroquant®, Supelco®
: Merck
VWR® Kjeldahl Tablets
: VWR Chemicals
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test kits, chlorine, Spectroquant®, Supelco®
: Merck
Formalin 10%, Q Path®, buffered
: VWR Chemicals
Papanicolaou's solution 2b Orange II solution stain for cytology, Sigma-Aldrich®
: Merck
Test kits, nitrate, Spectroquant®, Supelco®
: Merck
May Grunwald's eosin-methylene blue modified solution stain for hematology, for cytology, Sigma-Aldrich®
: Merck
Test kits, chloride, Spectroquant®, Supelco®
: Merck