Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SONIDEGIB (FREE BASE) 1 * 10 mg
Catalog # ENZOENZCHM2380010
Supplier:
Enzo Life Sciences
Expand image
SONIDEGIB (FREE BASE) 1 * 10 mg
Catalog # ENZOENZCHM2380010
Supplier:
Enzo Life Sciences
Frequently Bought Together
20G PL SILICA HR 1 (142 TESTS) 1 * 20 g
Supplier: WATER ID PRIMELAB
Bromophenol blue
Supplier: Hach
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Anti-Cdc25A (phospho S18) Rabbit Recombinant Multiclonal Antibody [clone: RP23040112]
Supplier: Abcam
Potassium hydroxide 8 mol/l (8 N) in aqueous solution
Supplier: Hach
Anti-KAT1 / HAT1 Rabbit Recombinant Multiclonal Antibody [clone: RP23040271]
Supplier: Abcam