Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RUCAPARIB PHOSPHATE 1 * 200 mg
ENZOENZCHM2360200
:
ENZO LIFE SCIENCES
RUCAPARIB PHOSPHATE 1 * 200 mg
ENZOENZCHM2360200
:
ENZO LIFE SCIENCES
Frequently Bought Together
Sodium hydrogen carbonate ≥98%, TECHNICAL
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Sodium hydrogen carbonate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
: VWR Chemicals
Disodium hydrogen phosphate dodecahydrate 98.5-102.5% (by acidimetry), crystals, EMSURE® Ph. Eur., BP, JP, USP, Supelco®
: Merck
Sodium acetate, anhydrous, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Ammonium heptamolybdate tetrahydrate ISO, ACS, Reag. Ph. Eur. analytical reagent, Supelco®
: Merck
Sodium acetate trihydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Sodium dihydrogen phosphate dihydrate, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
: Merck
Potassium chloride, EMSURE® for analysis, Supelco®
: Merck
Potassium dihydrogen phosphate, EMSURE® ISO for analysis, Supelco®
: Merck
Sodium sulphate, anhydrous, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Sodium chloride ≥98%, TECHNICAL
: VWR Chemicals
Water, EMSURE® for analysis, Supelco®
: Merck
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Magnesium sulphate heptahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Sodium hydrogen carbonate 99.7-100.3%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals