Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RUCAPARIB (FREE BASE) 1 * 25 mg
Catalog # ENZOENZCHM2350025
Supplier:
Enzo Life Sciences
Expand image
RUCAPARIB (FREE BASE) 1 * 25 mg
Catalog # ENZOENZCHM2350025
Supplier:
Enzo Life Sciences
Frequently Bought Together
Potassium hydroxide 8 mol/l (8 N) in aqueous solution
Supplier: Hach
VWR® Powder Packs for Determination of Nitrite, Reagents for Photometers
Supplier: VWR Chemicals
SONIDEGIB (FREE BASE) 1 * 10 mg
Supplier: Enzo Life Sciences
Bromophenol blue
Supplier: Hach
AB 548820 4-[(4-Hydroxyphenyl)diazenyl] benzenesulfonic acid CAS 2918-83-4 1g 1 * 1 g
Supplier: abcr
Anti-KAT1 / HAT1 Rabbit Recombinant Multiclonal Antibody [clone: RP23040271]
Supplier: Abcam