NINTEDANIB . ETHANESULFONATE SALT 1 * 1 g
Katalogové číslo ENZOENZCHM2291000
Dodavatel: ENZO LIFE SCIENCES
NINTEDANIB . ETHANESULFONATE SALT 1 * 1 g
Katalogové číslo ENZOENZCHM2291000
Dodavatel: ENZO LIFE SCIENCES
Frequently Bought Together
Cesium chloride analytical reagent, Supelco®
Dodavatel: Merck
Sodium acetate trihydrate 99.0-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Silver sulphate ≥98%, TECHNICAL
Dodavatel: VWR Chemicals
Potassium nitrate ≥98%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Manganese(II) chloride tetrahydrate 99.0-101.0%, AnalaR NORMAPUR® ACS analytical reagent
Dodavatel: VWR Chemicals
Aluminium hydroxide, fine powder
Dodavatel: Merck
Disodium metasilicate pentahydrate, TECHNICAL
Dodavatel: VWR Chemicals
Sodium benzoate, Sigma-Aldrich®
Dodavatel: Merck
SULFATE PENTAHYDRATE 1 * 100 g
Dodavatel: Merck
Sodium perchlorate, anhydrous ACS
Dodavatel: Thermo Fisher Scientific
Magnesium sulphate heptahydrate 99.0-100.5% Ph. Eur.
Dodavatel: VWR Chemicals
Methyl orange ACS
Dodavatel: Thermo Fisher Scientific
Sodium hydrogen carbonate ≥99%
Dodavatel: Thermo Fisher Scientific
Potassium iodide 99+% for analysis
Dodavatel: Thermo Fisher Scientific