Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
NERATINIB (FREE BASE) 1 * 10 mg
Catalog # ENZOENZCHM2280010
Supplier:
Enzo Life Sciences
Expand image
NERATINIB (FREE BASE) 1 * 10 mg
Catalog # ENZOENZCHM2280010
Supplier:
Enzo Life Sciences
Frequently Bought Together
Reagents for molybdate determination using Lovibond® MD Photometers
Supplier: Lovibond
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
Supplier: Honeywell Chemicals
PALBOCICLIB (FREE BASE) 1 * 2 g
Supplier: ENZO LIFE SCIENCES
Nitrite VHR L, 500 ml
Supplier: Lovibond