0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
IDELALISIB (FREE BASE) 1 * 500 mg
ENZOENZCHM2240500
:
ENZO LIFE SCIENCES
IDELALISIB (FREE BASE) 1 * 500 mg
ENZOENZCHM2240500
:
ENZO LIFE SCIENCES
Frequently Bought Together
Control slides, ISOSLIDE®
: Merck
Reagents for Phosphate Determination using Lovibond® MD photometers
: Lovibond
Sulfuric Acid TS
: CUSTOM MADE CHEMICALS LAB
Pfbbr 5g 1 * 5 g
: Thermo Fisher Scientific
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
: Hach
12 components: 100mg/l each of Al Ba B Co Cr Fe P Mn Ni Pb Cu Zn in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 1 * 100 mL
: CUSTOM MADE CHEMICALS LAB