DACOMITINIB (FREE BASE) 1 * 500 mg
Catalog # ENZOENZCHM2210500
Supplier: Enzo Life Sciences
Frequently Bought Together
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
pH indicator test strips, pH-Fix
Supplier: MACHEREY-NAGEL
Eosin Y (yellowish) in aqueous solution, Q Path® for microscopy
Supplier: VWR Chemicals
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Reagents for chlorine determination using Lovibond® MD photometers
Supplier: Lovibond
VWR® Cryolab Q Path, Freezing Aerosol
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Supplier: VWR Chemicals
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Standards for 2100N and 2100N IS
Supplier: Hach