DACOMITINIB (FREE BASE) 1 * 250 mg
Catalog # ENZOENZCHM2210250
Supplier: Enzo Life Sciences
Frequently Bought Together
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Joint grease, K.W.S.
Supplier: POLLATH
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
pH indicator test strips, pH-Fix
Supplier: MACHEREY-NAGEL
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach